9-(2-Hydroxyethyl)adenine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-9-(2-hydroxyethyl)purine

A335038-25G A335038-500G A335038-5G A335038-100G
76817-210EA 39 USD
76817-210 76817-212 76817-214 77672-716
9-(2-Hydroxyethyl)adenine 98%
9-(2-Hydroxyethyl)adenine

Precaución: for Research Use Only

Formula: C₇H₉N₅O
MW: 179.18 g/mol
MDL Number: MFCD04220770
CAS Number: 707-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR