5,7-Dibromo-1H-indole-2,3-dione 95%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dibromoindoline-2,3-dione

A334336-1G A334336-250MG A334336-5G
76817-204EA 56.23 USD
76817-204 76817-206 76817-208
5,7-Dibromo-1H-indole-2,3-dione 95%
5,7-Dibromo-1H-indole-2,3-dione

Precaución: for Research Use Only

Formula: C₈H₃Br₂NO₂
MW: 304.94 g/mol
MDL Number: MFCD00034382
CAS Number: 6374-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR