(4-Aminophenyl)(4-methylpiperazin-1-yl)methanone 95%

Supplier: Ambeed
Warning

A333960-5G A333960-250MG
76817-196EA 381.93 USD
76817-196 77672-704
(4-Aminophenyl)(4-methylpiperazin-1-yl)methanone 95%
(4-Aminophenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₇N₃O
MW: 219.2800 g/mol
MDL Number: MFCD00973892
CAS Number: 55121-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR