(R)-Methyl 2-amino-2-cyclohexylacetate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: H-D-Chg-OMe·HCl

A333383-10G A333383-1G A333383-5G A333383-100G
76817-190EA 42 USD
76817-190 76817-192 76817-194 77883-915
(R)-Methyl 2-amino-2-cyclohexylacetate hydrochloride 95%
(R)-Methyl 2-amino-2-cyclohexylacetate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD08275848
CAS Number: 14328-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR