2-Hydroxy-6-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A332435-10G A332435-5G
76817-182EA 1380.24 USD
76817-182 76817-184
2-Hydroxy-6-nitrobenzoic acid 98%
2-Hydroxy-6-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₅
MW: 183.11 g/mol
MDL Number: MFCD00031281
CAS Number: 601-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR