tert-Butyl 4-(6-aminopyridin-3-yl)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A331079-5G A331079-1G A331079-250MG
76817-176EA 129 USD
76817-176 77593-954 77883-907
tert-Butyl 4-(6-aminopyridin-3-yl)piperidine-1-carboxylate 95%
tert-Butyl 4-(6-aminopyridin-3-yl)piperidine-1-carboxylate
Formula: C₁₅H₂₃N₃O₂
MDL Number: MFCD11111266
CAS Number: 1198408-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR