1-Benzhydrylazetidin-3-amine hydrochloride 95%

Supplier: Ambeed
Warning

A330626-1G A330626-5G
76817-172EA 8.49 USD
76817-172 76817-174
1-Benzhydrylazetidin-3-amine hydrochloride 95%
1-Benzhydrylazetidin-3-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₁₉ClN₂
MW: 274.78 g/mol
MDL Number: MFCD01665361
CAS Number: 1965305-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR