7-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine 98%
Supplier: Ambeed
|
Warning
|
A330563-100MG
A330563-10G
A330563-250MG
A330563-5G
A330563-1G
76817-164EA
8
USD
76817-164
76817-166
76817-168
76817-170
77593-952
7-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine 98%
7-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine
Precaución: for Research Use Only
Formula:
C₇H₆BrN₃ |
MDL Number:
MFCD12827554 CAS Number: 1190927-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...