5-Bromo-1-methyl-1H-pyrazol-3-amine 95%

Supplier: Ambeed
Warning

A330495-100MG A330495-1G A330495-250MG A330495-5G
76817-156EA 44.56 USD
76817-156 76817-158 76817-160 76817-162
5-Bromo-1-methyl-1H-pyrazol-3-amine 95%
5-Bromo-1-methyl-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₄H₆BrN₃
MW: 176.02 g/mol
MDL Number: MFCD16620114
CAS Number: 89088-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR