(R)-1-Methoxypropan-2-amine hydrochloride 95%

Supplier: Ambeed
Warning

A330216-100MG A330216-1G A330216-5G A330216-250MG
76817-150EA 10 USD
76817-150 76817-152 76817-154 77883-901
(R)-1-Methoxypropan-2-amine hydrochloride 95%
(R)-1-Methoxypropan-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₂ClNO
MDL Number: MFCD12405301
CAS Number: 626220-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR