N,N'-Dibenzylethylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: N1,N2-Dibenzylethane-1,2-diamine, DBED

A329988-100G A329988-25G A329988-5G
76817-140EA 121 USD
76817-140 76817-142 76817-144
N,N'-Dibenzylethylenediamine 98%
N,N'-Dibenzylethylenediamine

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
MDL Number: MFCD00004771
CAS Number: 140-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR