(4-(Methoxymethoxy)-3,5-dimethylphenyl)boronic acid 96%

Supplier: Ambeed
Warning

Synonyms: 4-(Methoxymethoxy)-3,5-dimethylphenylboronic acid

A326434-1G A326434-250MG
76817-114EA 81 USD
76817-114 76817-116
(4-(Methoxymethoxy)-3,5-dimethylphenyl)boronic acid 96%
(4-(Methoxymethoxy)-3,5-dimethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₄
MDL Number: MFCD11856041
CAS Number: 223128-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR