6-Bromo-1H-indole-3-carbonitrile 97%

Supplier: Ambeed
Warning

A326162-100MG A326162-10G A326162-250MG A326162-5G A326162-1G
76817-102EA 21.22 USD
76817-102 76817-104 76817-106 76817-108 77672-678
6-Bromo-1H-indole-3-carbonitrile 97%
6-Bromo-1H-indole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅BrN₂
MW: 221.05 g/mol
MDL Number: MFCD10700148
CAS Number: 224434-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR