Quinolin-4(1H)-one 95%

Supplier: Ambeed
Warning

A325402-100G A325402-10G A325402-25G A325402-5G
76817-094EA 88.06 USD
76817-094 76817-096 76817-098 76817-100
Quinolin-4(1H)-one 95%
Quinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD00006777
CAS Number: 529-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR