3-Amino-4-phenylbutanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: H-DL-β-HoPhe-OH, DL-β-Homophenylalanine

A324373-1G A324373-250MG A324373-5G
76817-084EA 56.23 USD
76817-084 76817-086 76817-088
3-Amino-4-phenylbutanoic acid 95%
3-Amino-4-phenylbutanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD02090626
CAS Number: 15099-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR