2-Bromo-6-ethylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-ethyl-pyridine

A323538-100MG A323538-1G
76817-080EA 33.95 USD
76817-080 77550-900
2-Bromo-6-ethylpyridine 95%
2-Bromo-6-ethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD11869660
CAS Number: 83004-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR