tert-Butyl 2-methyl-3-oxopiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A323032-1G A323032-250MG
76817-070EA 161.26 USD
76817-070 76817-072
tert-Butyl 2-methyl-3-oxopiperidine-1-carboxylate 95%
tert-Butyl 2-methyl-3-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
MDL Number: MFCD08445688
CAS Number: 741737-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR