4-Bromo-2,6-dichloroaniline 97%
Supplier: Ambeed
|
Warning
|
A321012-100G
A321012-10G
A321012-1G
A321012-25G
A321012-5G
76817-052EA
79.57
USD
76817-052
76817-054
76817-056
76817-058
76817-060
4-Bromo-2,6-dichloroaniline 97%
4-Bromo-2,6-dichloroaniline
Precaución: for Research Use Only
Formula:
C₆H₄BrCl₂N MW: 240.91 g/mol |
MDL Number:
MFCD00025162 CAS Number: 697-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...