6-Fluoro-5-iodopyridin-2-amine 98%

Supplier: Ambeed
Warning

A319927-1G A319927-250MG A319927-5G
76817-032EA 55.17 USD
76817-032 76817-034 76817-036
6-Fluoro-5-iodopyridin-2-amine 98%
6-Fluoro-5-iodopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄FIN₂
MW: 238.002 g/mol
MDL Number: MFCD09032812
CAS Number: 884660-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR