3,5-Diiodopyridine 95%

Supplier: Ambeed
Warning

A318813-100MG A318813-250MG A318813-1G
76817-024EA 116.7 USD
76817-024 76817-026 77672-638
3,5-Diiodopyridine 95%
3,5-Diiodopyridine

Precaución: for Research Use Only

Formula: C₅H₃I₂N
MW: 330.893 g/mol
MDL Number: MFCD01928488
CAS Number: 53710-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR