5-Bromo-2-chlorobenzene-1-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A318641-1G A318641-250MG A318641-5G
76817-018EA 35 USD
76817-018 76817-020 76817-022
5-Bromo-2-chlorobenzene-1-sulfonyl chloride 95%
5-Bromo-2-chlorobenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂O₂S
MDL Number: MFCD12026193
CAS Number: 81226-68-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR