Methyl 2-fluoro-5-hydroxybenzoate 97%
Supplier: Ambeed
|
Warning
|
A318609-10G
A318609-1G
A318609-250MG
A318609-5G
A318609-25G
76817-010EA
143
USD
76817-010
76817-012
76817-014
76817-016
77883-863
Methyl 2-fluoro-5-hydroxybenzoate 97%
Methyl 2-fluoro-5-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₈H₇FO₃ |
MDL Number:
MFCD12406843 CAS Number: 1084801-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...