trans-2-(4-(4-(4-Amino-7,7-dimethyl-7H-pyrimido[4,5-b][1,4]oxazin-6-yl)phenyl)cyclohexyl)acetic acid 98+%

Supplier: Ambeed
Warning

Synonyms: T863

A318437-10MG A318437-1MG A318437-25MG A318437-5MG
76816-998EA 47 USD
76816-998 76817-000 76817-002 76817-004
trans-2-(4-(4-(4-Amino-7,7-dimethyl-7H-pyrimido[4,5-b][1,4]oxazin-6-yl)phenyl)cyclohexyl)acetic acid 98+%
trans-2-(4-(4-(4-Amino-7,7-dimethyl-7H-pyrimido[4,5-b][1,4]oxazin-6-yl)phenyl)cyclohexyl)acetic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₆N₄O₃
MDL Number: MFCD11977270
CAS Number: 701232-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR