4-Bromo-2-hydrazinopyridine 97%

Supplier: Ambeed
Danger

Synonyms: 4-Bromo-2-hydrazinylpyridine

A318147-1G A318147-250MG
76816-994EA 106.09 USD
76816-994 76816-996
4-Bromo-2-hydrazinopyridine 97%
4-Bromo-2-hydrazinopyridine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MW: 188.03 g/mol
MDL Number: MFCD00234091
CAS Number: 1019918-39-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR