6-Chloro-4-methylnicotinonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-cyano-4-methylpyridine, 6-Chloro-4-methylpyridine-3-carbonitrile

A318002-100MG A318002-1G A318002-250MG
76816-988EA 67.9 USD
76816-988 76816-990 76816-992
6-Chloro-4-methylnicotinonitrile 95%
6-Chloro-4-methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.581 g/mol
MDL Number: MFCD17214204
CAS Number: 66909-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR