6-Amino-1,3-dimethyluracil 97%

Supplier: Ambeed
Warning

A316401-1000G A316401-100G A316401-10G A316401-25G A316401-500G
76816-964EA 37 USD
76816-964 76816-966 76816-968 76816-970 76816-972 76816-974
6-Amino-1,3-dimethyluracil 97%
6-Amino-1,3-dimethyluracil

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MDL Number: MFCD00006552
CAS Number: 6642-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR