Ethyl 4-ethoxypicolinate 97%

Supplier: Ambeed
Warning

A314941-100MG A314941-1G A314941-250MG
76816-924EA 70 USD
76816-924 76816-926 76816-928
Ethyl 4-ethoxypicolinate 97%
Ethyl 4-ethoxypicolinate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
MDL Number: MFCD13181658
CAS Number: 71777-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR