DL-Glutamic acid 97%

Supplier: Ambeed
Warning

A314874-1000G A314874-100G A314874-25G A314874-500G
76816-916EA 61 USD
76816-916 76816-918 76816-920 76816-922
DL-Glutamic acid 97%
DL-Glutamic acid

Precaución: for Research Use Only

Formula: C₅H₉NO₄
MDL Number: MFCD00063113
CAS Number: 617-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR