2-Chloro-3-methylaniline 97%

Supplier: Ambeed
Warning

A314133-1G A314133-250MG A314133-5G
76816-904EA 33.95 USD
76816-904 76816-906 76816-908
2-Chloro-3-methylaniline 97%
2-Chloro-3-methylaniline

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.598 g/mol
MDL Number: MFCD11113332
CAS Number: 29027-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR