(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: cis-1-Benzyl-N3,4-dimethylpiperidin-3-amine dihydrochloride, cis-1-Benzyl-4-methyl-3-(methylamino)piperidine dihydrochloride

A312475-100G A312475-25G A312475-5G
76816-900EA 347 USD
76816-900 76816-902 77883-850
(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride 95%
(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₄Cl₂N₂
MW: 291.26 g/mol
MDL Number: MFCD09864744
CAS Number: 1062580-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR