Sodium 2-amino-9-((2-hydroxyethoxy)methyl)-9H-purin-6-olate 98%

Supplier: Ambeed
Danger

Synonyms: Acyclovir sodium, Sodium 2-((2-amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethanolate

A311430-5G A311430-25G
76816-898EA 20.16 USD
76816-898 77672-590
Sodium 2-amino-9-((2-hydroxyethoxy)methyl)-9H-purin-6-olate 98%
Sodium 2-amino-9-((2-hydroxyethoxy)methyl)-9H-purin-6-olate

Precaución: for Research Use Only

Formula: C₈H₁₀N₅NaO₃
MW: 247.19 g/mol
MDL Number: MFCD01694138
CAS Number: 69657-51-8
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR