6-Hydroxypyrimidine-4-carboxamide 98+%

Supplier: Ambeed
Warning

A309975-1G A309975-250MG
76816-890EA 111.4 USD
76816-890 77672-578
6-Hydroxypyrimidine-4-carboxamide 98+%
6-Hydroxypyrimidine-4-carboxamide

Precaución: for Research Use Only

Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
MDL Number: MFCD21340312
CAS Number: 98024-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR