Methyl 4-(2-bromophenyl)-2,4-dioxobutanoate 97%

Supplier: Ambeed
Warning

A309479-1G A309479-250MG
76816-886EA 182.48 USD
76816-886 76816-888
Methyl 4-(2-bromophenyl)-2,4-dioxobutanoate 97%
Methyl 4-(2-bromophenyl)-2,4-dioxobutanoate

Precaución: for Research Use Only

Formula: C₁₁H₉BrO₄
MW: 285.09 g/mol
MDL Number: MFCD11188864
CAS Number: 1035235-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR