1H-Indole-6-acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(1H-Indol-6-yl)acetic acid

A308491-1G A308491-250MG
76816-866EA 165.51 USD
76816-866 76816-868
1H-Indole-6-acetic acid 95%
1H-Indole-6-acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
MDL Number: MFCD09837606
CAS Number: 39689-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR