2,4-Dichloro-6-fluorobenzaldehyde 95%

Supplier: Ambeed
Warning

A307380-1G A307380-250MG A307380-5G
76816-834EA 30 USD
76816-834 76816-836 77883-837
2,4-Dichloro-6-fluorobenzaldehyde 95%
2,4-Dichloro-6-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO
MDL Number: MFCD12404935
CAS Number: 681435-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR