Ethyl-2-chloroacetoacetate 98%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-chloroacetoacetate

A304485-25G A304485-100G
76816-818EA 10 USD
76816-818 77883-835
Ethyl-2-chloroacetoacetate 98%
Ethyl-2-chloroacetoacetate

Precaución: for Research Use Only

Formula: C₆H₉ClO₃
MDL Number: MFCD00009141
CAS Number: 609-15-4
UN: 2920
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR