(R)-2-Hydroxy-N,N-dimethyl-3-((2-((1-(5-methylfuran-2-yl)propyl)amino)-3,4-dioxocyclobut-1-en-1-yl)amino)benzamide ≥98%

Supplier: Ambeed
Danger

Synonyms: Navarixin

A303940-10MG A303940-1MG A303940-50MG A303940-5MG
76816-802EA 88.06 USD
76816-802 76816-804 76816-806 76816-808
(R)-2-Hydroxy-N,N-dimethyl-3-((2-((1-(5-methylfuran-2-yl)propyl)amino)-3,4-dioxocyclobut-1-en-1-yl)amino)benzamide ≥98%
(R)-2-Hydroxy-N,N-dimethyl-3-((2-((1-(5-methylfuran-2-yl)propyl)amino)-3,4-dioxocyclobut-1-en-1-yl)amino)benzamide

Precaución: for Research Use Only

Formula: C₂₁H₂₃N₃O₅
MW: 397.42 g/mol
MDL Number: MFCD16628072
CAS Number: 473727-83-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR