4-Bromo-2-ethylbenzaldehyde 95%

Supplier: Ambeed
Warning

A301708-1G A301708-250MG A301708-5G
76816-784EA 196 USD
76816-784 76816-786 76816-788
4-Bromo-2-ethylbenzaldehyde 95%
4-Bromo-2-ethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO
MDL Number: MFCD09835094
CAS Number: 1114808-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR