Methyl 3-aminobenzofuran-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-2-benzo[b]furancarboxylate

A301059-1G A301059-250MG A301059-5G
76816-766EA 60.48 USD
76816-766 76816-768 76816-770
Methyl 3-aminobenzofuran-2-carboxylate 98%
Methyl 3-aminobenzofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MW: 191.183 g/mol
MDL Number: MFCD00466275
CAS Number: 57805-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR