(2-Fluoro-4-hydroxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A300876-1G A300876-250MG A300876-5G
76816-760EA 90 USD
76816-760 76816-762 76816-764
(2-Fluoro-4-hydroxyphenyl)boronic acid 97%
(2-Fluoro-4-hydroxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₆BFO₃
MDL Number: MFCD11101444
CAS Number: 1376989-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR