Ethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate hydrochloride

A299941-100MG A299941-250MG A299941-1G
76816-750EA 39.26 USD
76816-750 76816-752 77612-726
Ethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate hydrochloride 95%
Ethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄ClN₃O₂
MW: 231.6794 g/mol
MDL Number: MFCD09259961
CAS Number: 623906-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR