N-Boc-D-prolinol 97%

Supplier: Ambeed
Warning

A299194-100G A299194-10G A299194-1G A299194-25G A299194-500G A299194-5G
76816-728EA 103 USD
76816-728 76816-730 76816-732 76816-734 76816-736 76816-738
N-Boc-D-prolinol 97%
N-Boc-D-prolinol

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD00040580
CAS Number: 83435-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR