5-Bromo-1,2,3-triazine 97%

Supplier: Ambeed
Warning

A299113-250MG A299113-100MG A299113-1G
76816-722EA 66 USD
76816-722 77883-813 77883-814
5-Bromo-1,2,3-triazine 97%
5-Bromo-1,2,3-triazine

Precaución: for Research Use Only

Formula: C₃H₂BrN₃
MDL Number: MFCD13193660
CAS Number: 114078-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR