Methyl 3-bromo-2-methyl-5-nitrobenzoate 97%

Supplier: Ambeed
Warning

A297434-10G A297434-1G A297434-250MG A297434-25G A297434-5G
76816-684EA 76 USD
76816-684 76816-686 76816-688 76816-690 76816-692
Methyl 3-bromo-2-methyl-5-nitrobenzoate 97%
Methyl 3-bromo-2-methyl-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrNO₄
MDL Number: MFCD08273772
CAS Number: 885519-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR