3-Bromo-5-fluorophenylacetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Bromo-5-fluorophenyl)acetic acid

A296990-1G A296990-250MG A296990-5G A296990-25G
76816-678EA 24 USD
76816-678 76816-680 76816-682 77881-819
3-Bromo-5-fluorophenylacetic acid 95%
3-Bromo-5-fluorophenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MDL Number: MFCD09258961
CAS Number: 202000-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR