4-Bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A296012-100MG A296012-1G A296012-1MG A296012-250MG A296012-5G
76816-668EA 27.59 USD
76816-668 76816-670 76816-672 76816-674 76816-676
4-Bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine 95%
4-Bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.48 g/mol
MDL Number: MFCD12828064
CAS Number: 916176-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR