Urocanic acid 95%

Supplier: Ambeed
Warning

A295948-100MG A295948-1G A295948-1MG A295948-250MG A295948-5G A295948-100G
76816-658EA 6 USD
76816-658 76816-660 76816-662 76816-664 76816-666 77883-803
Urocanic acid 95%
Urocanic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MDL Number: MFCD00005203
CAS Number: 104-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR