4-(Hydroxyimino)methylphenylboronic acid 95%

Supplier: Ambeed
Danger

Synonyms: (4-((Hydroxyimino)methyl)phenyl)boronic acid

A293410-1G A293410-250MG A293410-5G
76816-614EA 42.44 USD
76816-614 76816-616 76816-618
4-(Hydroxyimino)methylphenylboronic acid 95%
4-(Hydroxyimino)methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₃
MW: 164.96 g/mol
MDL Number: MFCD04039325
CAS Number: 179942-51-9
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR