4-Hydroxy-2,6-dimethylnicotinic acid 95%

Supplier: Ambeed
Warning

A292093-250MG A292093-100MG A292093-1G
76816-604EA 37 USD
76816-604 77612-682 77883-789
4-Hydroxy-2,6-dimethylnicotinic acid 95%
4-Hydroxy-2,6-dimethylnicotinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD18258494
CAS Number: 33259-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR