2-Fluoro-5-methyl-3-pyridineboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-methylpyridine-3-boronic acid pinacol ester, 2-Fluoro-5-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A291952-1G A291952-250MG A291952-5G
76816-598EA 55 USD
76816-598 76816-600 76816-602
2-Fluoro-5-methyl-3-pyridineboronic acid pinacol ester 95%
2-Fluoro-5-methyl-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
MDL Number: MFCD08063092
CAS Number: 1073371-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR